Gene Transcripts

Gene Symbol
MECOM
Entrez Gene ID
2122
Gene Name
MDS1 and EVI1 complex locus
Gene Aliases
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
Chromosome Location
Chr. 3: 169083499 - 169663781 on Build GRCh38
UniGene ID
Hs.744090
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105077.3 NP_001098547.3 8 1049
NM_001105078.3 NP_001098548.2 7 1040
NM_001163999.1 NP_001157471.1 7 987
NM_001164000.1 NP_001157472.1 7 1916
NM_001205194.1 NP_001192123.1 7 1344
NM_004991.3 NP_004982.2 8 1682
NM_005241.3 NP_005232.2 7 984
XM_005247213.3 XP_005247270.1 8 1691
XM_005247214.3 XP_005247271.1 8 1691
XM_005247215.3 XP_005247272.1 8 1687
XM_005247219.2 XP_005247276.1 7 1656
XM_005247220.2 XP_005247277.1 7 997
XM_005247221.2 XP_005247278.1 7 980
XM_005247223.2 XP_005247280.1 7 1653
XM_011512546.2 XP_011510848.1 7 1444
XM_017005874.1 XP_016861363.1 7 2359
XM_017005875.1 XP_016861364.1 7 994
XM_017005876.1 XP_016861365.1 7 1656
GenBank mRNA AK292865.1 - 7 967
AK304098.1 - 8 1628
AK308404.1 - 8 1327
AK310509.1 - 6 828
BC130520.1 - 8 1023 40146551
BC143951.1 - 8 1023 9052469
BC143952.1 - 7 961 9052470
BX640908.1 - 7 1916
BX647613.1 - 7 1648
GQ352634.1 - 7 1248
X54989.1 - 7 982

View Full Product Details