Gene Transcripts

Gene Symbol
SLC30A4
Entrez Gene ID
7782
Gene Name
solute carrier family 30 member 4
Gene Aliases
ZNT4, znT-4
Chromosome Location
Chr. 15: 45479613 - 45523685 on Build GRCh38
UniGene ID
Hs.162989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013309.5 NP_037441.2 6, 7 1320
XM_011521997.2 XP_011520299.1 6, 7 1824
XM_017022560.1 XP_016878049.1 5, 6 1126
GenBank mRNA AF025409.1 - 5, 6 1008
AK290874.1 - 5, 6 1126
AY029482.1 - 2, 3 285
BC026089.2 - 6, 7 1320 4829839

View Full Product Details