Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003373.3 | NP_003364.1 | 6 | 766 | |
NM_014000.2 | NP_054706.1 | 6 | 766 | ||
GenBank mRNA | AK126978.1 | - | 6 | 1021 | |
AK304823.1 | - | 5 | 695 | ||
BC039174.1 | - | 6 | 753 | 4520338 | |
BX537994.1 | - | 8 | 631 | ||
EU794673.1 | - | 6 | 766 | ||
M33308.1 | - | 6 | 722 |