Gene Transcripts
- Gene Symbol
- NUP54
- Entrez Gene ID
- 53371
- Gene Name
- nucleoporin 54
- Gene Aliases
- Chromosome Location
- Chr. 4: 76114659 - 76148515 on Build GRCh38
- UniGene ID
- Hs.430435
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278603.1 |
NP_001265532.1
|
9, 10 |
1289 |
|
NM_017426.3 |
NP_059122.2
|
10, 11 |
1433 |
|
NR_103780.1 |
|
9, 10 |
1206 |
|
NR_103781.1 |
|
8, 9 |
1098 |
|
NR_103782.1 |
|
10, 11 |
1203 |
|
XM_006714237.1 |
XP_006714300.1
|
11, 12 |
1410 |
|
XM_011532033.2 |
XP_011530335.1
|
9, 10 |
1268 |
|
XM_011532034.2 |
XP_011530336.1
|
9, 10 |
1676 |
|
XM_017008294.1 |
XP_016863783.1
|
10, 11 |
1820 |
|
XM_017008295.1 |
XP_016863784.1
|
9, 10 |
1593 |
|
GenBank mRNA |
AK001517.1 |
- |
10, 11 |
1315 |
|
AK222752.1 |
- |
10, 11 |
1320 |
|
AK223352.1 |
- |
10, 11 |
1316 |
|
AK300036.1 |
- |
9, 10 |
1171 |
|
AK307571.1 |
- |
10, 11 |
1093 |
|
AK315160.1 |
- |
10, 11 |
1313 |
|
BC012559.1 |
- |
10, 11 |
1330 |
|
BX538002.1 |
- |
8, 9 |
998 |
|
View Full Product Details