Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018358.2 | NP_060828.2 | 16 | 1629 | |
XM_011512959.2 | XP_011511261.1 | 16 | 1633 | ||
GenBank mRNA | AK002060.1 | - | 16 | 1606 | |
AK024758.1 | - | 10 | 828 | ||
AK027288.1 | - | 10 | 897 | ||
AK092415.1 | - | 16 | 1512 | ||
AK097073.1 | - | 15 | 1896 | ||
AK126150.1 | - | 2 | 1274 | ||
AK290106.1 | - | 16 | 1501 | ||
AK299433.1 | - | 16 | 1393 | ||
BC009253.2 | - | 16 | 1508 | 3028775 | |
BC035652.1 | - | 16 | 1503 | ||
BC051754.2 | - | 16 | 1503 | 6064628 | |
BC051884.1 | - | 16 | 1503 | 5105761 | |
KU178694.1 | - | 16 | 1496 | ||
KU178695.1 | - | 16 | 1453 |