Gene Transcripts

Gene Symbol
SLCO1B3
Entrez Gene ID
28234
Gene Name
solute carrier organic anion transporter family member 1B3
Gene Aliases
HBLRR, LST-2, LST-3TM13, LST3, OATP-8, OATP1B3, OATP8, SLC21A8
Chromosome Location
Chr. 12: 20810704 - 20916911 on Build GRCh38
UniGene ID
Hs.504966
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019844.3 NP_062818.1 13 1900
GenBank mRNA AF187815.1 - 11 1659
AJ251506.1 - 11 1703
AK055874.1 - 13 1878
AY342017.1 - 11 1673

View Full Product Details