Gene Transcripts
- Gene Symbol
- EDA2R
- Entrez Gene ID
- 60401
- Gene Name
- ectodysplasin A2 receptor
- Gene Aliases
- EDA-A2R, EDAA2R, TNFRSF27, XEDAR
- Chromosome Location
- Chr. X: 66594384 - 66639303 on Build GRCh38
- UniGene ID
- Hs.302017
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199687.2 |
NP_001186616.1
|
3, 4 |
382 |
|
NM_001242310.1 |
NP_001229239.1
|
2, 3 |
284 |
|
NM_001324206.1 |
NP_001311135.1
|
3, 4 |
384 |
|
NM_021783.4 |
NP_068555.1
|
3, 4 |
384 |
|
NR_136726.1 |
|
3, 4 |
382 |
|
NR_136727.1 |
|
3, 4 |
384 |
|
XM_011531000.1 |
XP_011529302.1
|
3, 4 |
334 |
|
XM_011531001.1 |
XP_011529303.1
|
3, 4 |
382 |
|
XM_011531002.1 |
XP_011529304.1
|
3, 4 |
334 |
|
XM_011531004.1 |
XP_011529306.1
|
3, 4 |
334 |
|
XM_011531006.2 |
XP_011529308.1
|
3, 4 |
374 |
|
XM_011531007.2 |
XP_011529309.1
|
3, 4 |
374 |
|
XM_017029704.1 |
XP_016885193.1
|
3, 4 |
334 |
|
XM_017029706.1 |
XP_016885195.1
|
3, 4 |
334 |
|
GenBank mRNA |
AF298812.1 |
- |
2, 3 |
274 |
|
AK296264.1 |
- |
3, 4 |
334 |
|
AK314881.1 |
- |
3, 4 |
350 |
|
AY152724.1 |
- |
2, 3 |
274 |
|
AY358735.1 |
- |
2, 3 |
277 |
|
AY358736.1 |
- |
2, 3 |
277 |
|
BC034919.1 |
- |
3, 4 |
331 |
5434545 |
BP249106.1 |
- |
3, 4 |
350 |
|
DA215848.1 |
- |
3, 4 |
383 |
|
DA574050.1 |
- |
3, 4 |
334 |
|
View Full Product Details