Gene Transcripts

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
Chromosome Location
Chr. 4: 107931549 - 107953461 on Build GRCh38
UniGene ID
Hs.109087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183075.2 NP_898898.1 5 2918
XM_005262717.2 XP_005262774.1 5 2984
XM_005262720.2 XP_005262777.1 4 2294
GenBank mRNA AK026498.1 - 1 943
AY343323.1 - 5 2874
BC132767.1 - 5 2883 40146789
BC136483.1 - 5 2883 9020472

View Full Product Details