Gene Transcripts

Gene Symbol
FTCD
Entrez Gene ID
10841
Gene Name
formimidoyltransferase cyclodeaminase
Gene Aliases
LCHC1
Chromosome Location
Chr. 21: 46135981 - 46156481 on Build GRCh38
UniGene ID
Hs.415846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006657.2 NP_006648.1 13, 14 1576
NM_206965.1 NP_996848.1 13, 14 1576
XM_006723961.3 XP_006724024.2 13, 14 1996
XM_011529437.2 XP_011527739.1 13, 14 1998
GenBank mRNA AF169017.1 - 13, 14 1576
AF289021.1 - 13, 14 1576
AF289023.1 - 12, 13 1532
AF289024.1 - 13, 14 1601
BC032037.1 - 3, 4 863
BC052248.1 - 13, 14 1572 6270063
BC136383.1 - 13, 14 1568 9020370
BC136395.1 - 13, 14 1568 9020382
BI966164.1 - 4, 5 414
BM021970.1 - 4, 5 411
U91541.1 - 4, 5 347

View Full Product Details