Gene Transcripts
- Gene Symbol
- SAT2
- Entrez Gene ID
- 112483
- Gene Name
- spermidine/spermine N1-acetyltransferase family member 2
- Gene Aliases
- SSAT2
- Chromosome Location
- Chr. 17: 7626234 - 7627878 on Build GRCh38
- UniGene ID
- Hs.10846
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320845.1 |
NP_001307774.1
|
1, 2 |
335 |
|
NM_001320846.1 |
NP_001307775.1
|
1, 2 |
335 |
|
NM_001320847.1 |
NP_001307776.1
|
1, 2 |
318 |
|
NM_133491.4 |
NP_597998.1
|
1, 2 |
318 |
|
XM_017024073.1 |
XP_016879562.1
|
1, 2 |
335 |
|
XM_017024074.1 |
XP_016879563.1
|
1, 2 |
337 |
|
GenBank mRNA |
AF348524.1 |
- |
1, 2 |
77 |
|
BC011751.2 |
- |
1, 2 |
288 |
3534187 |
BC065922.1 |
- |
1 |
393 |
|
BC095466.1 |
- |
1, 2 |
273 |
|
BG748008.1 |
- |
1, 2 |
245 |
|
BP317593.1 |
- |
1, 2 |
336 |
|
HY155008.1 |
- |
1, 2 |
321 |
|
View Full Product Details