Gene Transcripts

Gene Symbol
MMP19
Entrez Gene ID
4327
Gene Name
matrix metallopeptidase 19
Gene Aliases
CODA, MMP18, RASI-1
Chromosome Location
Chr. 12: 55835430 - 55842983 on Build GRCh38
UniGene ID
Hs.591033
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272101.1 NP_001259030.1 6 1029
NM_002429.5 NP_002420.1 7 1169
NR_073606.1 7 1246
XM_006719401.3 XP_006719464.1 5 810
XM_011538359.1 XP_011536661.1 6 909
XM_017019308.1 XP_016874797.1 7 1261
XM_017019309.1 XP_016874798.1 7 1155
GenBank mRNA AK303202.1 - 6 982
BC050368.1 - 7 1108 5761074
BI761698.1 - 5 759
U37791.1 - 7 1097
U38320.1 - 3 1357
U38321.1 - 7 1092
U38322.1 - 3 903
U38431.1 - 7 1167
X92521.1 - 7 1117
Y08622.1 - 7 1116

View Full Product Details