Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001207024.1 | NP_001193953.1 | 2 | 397 | |
NM_002355.3 | NP_002346.1 | 2 | 397 | ||
XM_005253376.2 | XP_005253433.1 | 2 | 259 | ||
GenBank mRNA | AK222792.1 | - | 2 | 278 | |
AK291143.1 | - | 2 | 243 | ||
AK312689.1 | - | 2 | 282 | ||
BC024206.1 | - | 2 | 289 | 3530887 | |
BU166950.1 | - | 2 | 292 | ||
CR457412.1 | - | 1 | 122 | ||
DC341018.1 | - | 2 | 397 | ||
M16985.1 | - | 2 | 267 |