Gene Transcripts

Gene Symbol
MMP27
Entrez Gene ID
64066
Gene Name
matrix metallopeptidase 27
Gene Aliases
MMP-27
Chromosome Location
Chr. 11: 102690943 - 102705785 on Build GRCh38
UniGene ID
Hs.534479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022122.2 NP_071405.2 2 189
XM_011542948.2 XP_011541250.1 2 237
XM_011542949.2 XP_011541251.1 2 237
XM_017018120.1 XP_016873609.1 2 237
GenBank mRNA AF195192.1 - 2 172
AY358752.1 - 2 189

View Full Product Details