Gene Transcripts

Gene Symbol
SCGB2A1
Entrez Gene ID
4246
Gene Name
secretoglobin family 2A member 1
Gene Aliases
LPHC, LPNC, MGB2, UGB3
Chromosome Location
Chr. 11: 62208668 - 62213939 on Build GRCh38
UniGene ID
Hs.97644
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002407.2 NP_002398.1 2 150
GenBank mRNA AF071219.1 - 2 150
AJ224173.1 - 2 126
BC062218.1 - 2 149 30351353

View Full Product Details