Gene Transcripts

Gene Symbol
POU2F1
Entrez Gene ID
5451
Gene Name
POU class 2 homeobox 1
Gene Aliases
OCT1, OTF1, oct-1B
Chromosome Location
Chr. 1: 167220829 - 167427345 on Build GRCh38
UniGene ID
Hs.283402
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198783.1 NP_001185712.1 3 347
NM_001198786.1 NP_001185715.1 3 214
NM_002697.3 NP_002688.3 3 214
NR_037163.1 4 258
XM_011509653.2 XP_011507955.1 4 166
XM_011509654.2 XP_011507956.1 3 127
XM_011509655.1 XP_011507957.1 4 166
XM_017001508.1 XP_016856997.1 4 166
GenBank mRNA AK091438.1 - 5 642
AK302525.1 - 4 253
AK303201.1 - 3 152
AK308004.1 - 4 253
AY113189.1 - 3 166
BC001664.2 - 3 130 2966289
BC003571.1 - 3 130 3544378
BC052274.1 - 4 238 4622256
BX954888.1 - 3 215
DA937884.1 - 3 347
DC370634.1 - 4 258
KT438684.1 - 5 325
X13403.1 - 3 135

View Full Product Details