Gene Transcripts

Gene Symbol
NEUROG1
Entrez Gene ID
4762
Gene Name
neurogenin 1
Gene Aliases
AKA, Math4C, NEUROD3, bHLHa6, ngn1
Chromosome Location
Chr. 5: 135534282 - 135535949 on Build GRCh38
UniGene ID
Hs.248149
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006161.2 NP_006152.2 1 1080
GenBank mRNA AB593107.1 - 1 1092
BC008687.1 - 1 1080 3872172
BC028226.1 - 1 1080 5241507

View Full Product Details