Gene Transcripts

Gene Symbol
HOXA7
Entrez Gene ID
3204
Gene Name
homeobox A7
Gene Aliases
ANTP, HOX1, HOX1.1, HOX1A
Chromosome Location
Chr. 7: 27153716 - 27156677 on Build GRCh38
UniGene ID
Hs.610216
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006896.3 NP_008827.2 1 289
GenBank mRNA AF026397.1 - 1 289

View Full Product Details