Gene Transcripts

Gene Symbol
INPP5E
Entrez Gene ID
56623
Gene Name
inositol polyphosphate-5-phosphatase E
Gene Aliases
CORS1, CPD4, JBTS1, MORMS, PPI5PIV
Chromosome Location
Chr. 9: 136428615 - 136439861 on Build GRCh38
UniGene ID
Hs.120998
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318502.1 NP_001305431.1 10 2410
NM_019892.5 NP_063945.2 10 2413
XM_017014926.1 XP_016870415.1 10 2405
GenBank mRNA AF187891.1 - 10 2114
AL832846.1 - 10 2411
BC028032.1 - 10 2395 5242186
U45974.1 - 4 619

View Full Product Details