Gene Transcripts

Gene Symbol
HOXB4
Entrez Gene ID
3214
Gene Name
homeobox B4
Gene Aliases
HOX-2.6, HOX2, HOX2F
Chromosome Location
Chr. 17: 48575507 - 48578381 on Build GRCh38
UniGene ID
Hs.664706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024015.4 NP_076920.1 2 1160
GenBank mRNA AL137449.1 - 2 1084
BC049204.1 - 2 1160 5533346

View Full Product Details