Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002422.4 | NP_002413.1 | 7 | 1198 | |
GenBank mRNA | AK223283.1 | - | 7 | 1116 | |
AK223291.1 | - | 7 | 1117 | ||
AK313310.1 | - | 7 | 1114 | ||
BC069676.1 | - | 7 | 1118 | 7262578 | |
BC069716.1 | - | 7 | 1118 | 7262590 | |
BC074815.2 | - | 7 | 1100 | 30915319 | |
BC074869.2 | - | 7 | 1100 | 30915468 | |
BC105954.1 | - | 7 | 1111 | 40033171 | |
BC107490.1 | - | 7 | 1111 | 40033168 | |
BC107491.1 | - | 7 | 1111 | 40033170 | |
J03209.1 | - | 7 | 1055 | ||
X05232.1 | - | 7 | 1098 |