Gene Transcripts
- Gene Symbol
- TRPM6
- Entrez Gene ID
- 140803
- Gene Name
- transient receptor potential cation channel subfamily M member 6
- Gene Aliases
- CHAK2, HMGX, HOMG, HOMG1, HSH
- Chromosome Location
- Chr. 9: 74722495 - 74888094 on Build GRCh38
- UniGene ID
- Hs.272225
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001177310.1 |
NP_001170781.1
|
3, 4 |
217 |
|
NM_001177311.1 |
NP_001170782.1
|
3, 4 |
244 |
|
NM_017662.4 |
NP_060132.3
|
3, 4 |
398 |
|
XM_011518244.1 |
XP_011516546.1
|
3, 4 |
398 |
|
XM_011518245.1 |
XP_011516547.1
|
3, 4 |
398 |
|
XM_011518246.1 |
XP_011516548.1
|
3, 4 |
398 |
|
XM_011518247.1 |
XP_011516549.1
|
3, 4 |
398 |
|
XM_011518248.1 |
XP_011516550.1
|
3, 4 |
398 |
|
XM_011518249.1 |
XP_011516551.1
|
3, 4 |
398 |
|
XM_011518250.1 |
XP_011516552.1
|
3, 4 |
398 |
|
XM_011518252.2 |
XP_011516554.1
|
3, 4 |
398 |
|
XM_011518255.2 |
XP_011516557.1
|
3, 4 |
398 |
|
XM_017014287.1 |
XP_016869776.1
|
3, 4 |
398 |
|
XM_017014288.1 |
XP_016869777.1
|
3, 4 |
398 |
|
XM_017014289.1 |
XP_016869778.1
|
3, 4 |
398 |
|
GenBank mRNA |
AF350881.2 |
- |
3, 4 |
398 |
|
AF448232.1 |
- |
3, 4 |
160 |
|
AK057748.1 |
- |
3, 4 |
398 |
|
AY333282.1 |
- |
3, 4 |
223 |
|
AY333283.1 |
- |
3, 4 |
217 |
|
AY333284.1 |
- |
3, 4 |
244 |
|
AY333285.1 |
- |
3, 4 |
223 |
|
AY333286.1 |
- |
3, 4 |
223 |
|
AY333287.1 |
- |
3, 4 |
223 |
|
AY333288.1 |
- |
3, 4 |
223 |
|
View Full Product Details