Gene Transcripts

Gene Symbol
ZHX1
Entrez Gene ID
11244
Gene Name
zinc fingers and homeoboxes 1
Gene Aliases
Chromosome Location
Chr. 8: 123248450 - 123275541 on Build GRCh38
UniGene ID
Hs.521800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017926.2 NP_001017926.1 3 2381
NM_007222.4 NP_009153.3 3 2286
NR_037873.1 3 2217
GenBank mRNA AF106862.1 - 1 1700
AF195766.1 - 3 2049
AK289532.1 - 3 2217
BC040481.1 - 3 2106 5271344

View Full Product Details