Gene Transcripts

Gene Symbol
BTBD2
Entrez Gene ID
55643
Gene Name
BTB domain containing 2
Gene Aliases
Chromosome Location
Chr. 19: 1985438 - 2015703 on Build GRCh38
UniGene ID
Hs.465543
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017797.3 NP_060267.2 3 586
XM_005259593.1 XP_005259650.1 3 351
GenBank mRNA AF355797.1 - 3 586
AK225831.1 - 3 585
AY007116.1 - 3 308
BC069199.1 - 1 12
BC094820.1 - 3 320

View Full Product Details