Gene Transcripts

Gene Symbol
IKZF4
Entrez Gene ID
64375
Gene Name
IKAROS family zinc finger 4
Gene Aliases
EOS, ZNFN1A4
Chromosome Location
Chr. 12: 56007647 - 56038435 on Build GRCh38
UniGene ID
Hs.553221
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022465.3 NP_071910.3 4 971
XM_005269086.3 XP_005269143.1 6 513
XM_005269089.2 XP_005269146.1 4 615
XM_005269090.3 XP_005269147.1 5 419
XM_005269093.2 XP_005269150.1 3 890
XM_011538664.1 XP_011536966.1 6 516
XM_011538666.2 XP_011536968.1 5 364
XM_011538669.2 XP_011536971.1 4 314
XM_017019805.1 XP_016875294.1 8 761
XM_017019806.1 XP_016875295.1 8 1004
XM_017019807.1 XP_016875296.1 8 1001
XM_017019808.1 XP_016875297.1 7 686
XM_017019809.1 XP_016875298.1 8 758
XM_017019810.1 XP_016875299.1 8 856
XM_017019811.1 XP_016875300.1 7 683
XM_017019812.1 XP_016875301.1 6 759
XM_017019813.1 XP_016875302.1 6 756
XM_017019814.1 XP_016875303.1 5 665
XM_017019815.1 XP_016875304.1 5 662
XM_017019816.1 XP_016875305.1 5 422
GenBank mRNA AB058685.1 - 2 515
AF230809.1 - 3 223
AK122880.1 - 3 738
AK314543.1 - 3 336
BX647761.1 - 5 541

View Full Product Details