Gene Transcripts

Gene Symbol
ZNF34
Entrez Gene ID
80778
Gene Name
zinc finger protein 34
Gene Aliases
KOX32
Chromosome Location
Chr. 8: 144772224 - 144787371 on Build GRCh38
UniGene ID
Hs.631854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286769.1 NP_001273698.1 6 761
NM_001286770.1 NP_001273699.1 5 637
NM_030580.4 NP_085057.3 6 757
XM_011517314.2 XP_011515616.1 6 2084
XM_011517315.2 XP_011515617.1 6 1001
XM_011517316.2 XP_011515618.1 7 806
XM_011517318.2 XP_011515620.1 6 2084
XM_011517320.2 XP_011515622.1 5 667
XM_017013872.1 XP_016869361.1 7 807
XM_017013873.1 XP_016869362.1 6 747
XM_017013874.1 XP_016869363.1 6 781
XM_017013875.1 XP_016869364.1 7 947
XM_017013876.1 XP_016869365.1 5 621
GenBank mRNA AK096508.1 - 5 608
AL833814.1 - 6 741
BC004480.2 - 6 716 3938462
BC028136.1 - 3 859 5215496

View Full Product Details