Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033549.4 | NP_291027.3 | 6 | 3274 | |
NM_201627.2 | NP_963921.1 | 8 | 2334 | ||
NR_045218.1 | 7 | 2311 | |||
XM_006714929.2 | XP_006714992.1 | 7 | 2696 | ||
XM_006714930.2 | XP_006714993.1 | 7 | 2690 | ||
XM_006714931.3 | XP_006714994.1 | 8 | 2328 | ||
XM_006714932.3 | XP_006714995.1 | 8 | 2338 | ||
GenBank mRNA | AB063180.1 | - | 8 | 1360 | |
AB100366.1 | - | 6 | 2536 | ||
AB100367.1 | - | 8 | 1597 | ||
AF258579.1 | - | 2 | 1630 | ||
AK027601.1 | - | 8 | 2307 | ||
AK074052.1 | - | 2 | 5325 | ||
AL137593.1 | - | 4 | 1985 | ||
AL832960.1 | - | 5 | 2351 | ||
AX775789.1 | - | 6 | 2013 | ||
AX775791.1 | - | 6 | 2013 | ||
AX775793.1 | - | 8 | 2307 | ||
AX775797.1 | - | 6 | 2305 | ||
AX775799.1 | - | 6 | 3247 | ||
AX775801.1 | - | 5 | 3447 | ||
AX775803.1 | - | 5 | 2505 | ||
BC004956.1 | - | 7 | 1516 | ||
BC009762.2 | - | 2 | 1308 | ||
BC018765.1 | - | 6 | 2305 | ||
BC071887.1 | - | 7 | 1516 | ||
BC090953.1 | - | 6 | 2815 | 6017628 | |
BC111568.1 | - | 3 | 361 |