Gene Transcripts

Gene Symbol
PI4K2B
Entrez Gene ID
55300
Gene Name
phosphatidylinositol 4-kinase type 2 beta
Gene Aliases
PI4KIIB, PIK42B
Chromosome Location
Chr. 4: 25234031 - 25279209 on Build GRCh38
UniGene ID
Hs.191701
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018323.3 NP_060793.2 2 526
XM_005248174.1 XP_005248231.1 2 524
XM_005248175.4 XP_005248232.1 2 304
GenBank mRNA AF411320.1 - 2 393
AK001967.1 - 2 514
AK128622.1 - 3 431
AK296302.1 - 2 216
AY065990.1 - 2 504
AY091514.1 - 2 513
BC051749.1 - 2 526 6050442

View Full Product Details