Gene Transcripts
- Gene Symbol
- SLC5A10
- Entrez Gene ID
- 125206
- Gene Name
- solute carrier family 5 member 10
- Gene Aliases
- SGLT-5, SGLT5
- Chromosome Location
- Chr. 17: 18950676 - 19020691 on Build GRCh38
- UniGene ID
- Hs.462418
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042450.2 |
NP_001035915.1
|
14, 15 |
1730 |
|
NM_001270648.1 |
NP_001257577.1
|
13, 14 |
1649 |
|
NM_001270649.1 |
NP_001257578.1
|
13, 14 |
1622 |
|
NM_001282417.1 |
NP_001269346.1
|
14, 15 |
1839 |
|
NM_152351.4 |
NP_689564.3
|
14, 15 |
1778 |
|
XM_011523652.1 |
XP_011521954.1
|
15, 16 |
1997 |
|
XM_011523653.2 |
XP_011521955.1
|
14, 15 |
1969 |
|
XM_017024191.1 |
XP_016879680.1
|
15, 16 |
2046 |
|
GenBank mRNA |
AK057946.1 |
- |
14, 15 |
1779 |
|
AK298345.1 |
- |
13, 14 |
1608 |
|
BC034380.1 |
- |
14, 15 |
1675 |
|
BC039868.1 |
- |
14, 15 |
1856 |
4157703 |
BC062617.1 |
- |
13, 14 |
1603 |
|
View Full Product Details