Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318876.1 | NP_001305805.1 | 5, 6 | 589 | |
NM_203290.3 | NP_976035.1 | 5, 6 | 589 | ||
XM_011515000.2 | XP_011513302.1 | 5, 6 | 530 | ||
GenBank mRNA | AF008442.1 | - | 5, 6 | 515 | |
AF047441.1 | - | 5, 6 | 525 | ||
AK293561.1 | - | 4, 5 | 742 | ||
BC008118.1 | - | 5, 6 | 480 | ||
BC008863.2 | - | 5, 6 | 515 | 4128104 |