Gene Transcripts

Gene Symbol
DDR2
Entrez Gene ID
4921
Gene Name
discoidin domain receptor tyrosine kinase 2
Gene Aliases
MIG20a, NTRKR3, TKT, TYRO10
Chromosome Location
Chr. 1: 162631213 - 162783524 on Build GRCh38
UniGene ID
Hs.275757
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014796.1 NP_001014796.1 16 2375
NM_006182.2 NP_006173.2 15 2295
XM_006711344.3 XP_006711407.1 15 2336
XM_011509586.2 XP_011507888.1 15 2268
XM_011509587.2 XP_011507889.1 16 2329
GenBank mRNA AK095975.1 - 16 2348
AK225063.1 - 15 2214
AK314388.1 - 15 2244
BC052998.2 - 16 2346 6162716
BX640899.1 - 16 2332
X74764.1 - 15 2263

View Full Product Details