Gene Transcripts

Gene Symbol
KIF3A
Entrez Gene ID
11127
Gene Name
kinesin family member 3A
Gene Aliases
FLA10, KLP-20
Chromosome Location
Chr. 5: 132688808 - 132737633 on Build GRCh38
UniGene ID
Hs.43670
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300791.1 NP_001287720.1 2 414
NM_001300792.1 NP_001287721.1 2 414
NM_007054.6 NP_008985.3 2 414
XM_006714526.3 XP_006714589.1 2 469
XM_017008995.1 XP_016864484.1 2 469
XM_017008996.1 XP_016864485.1 2 469
GenBank mRNA AB209780.1 - 2 362
AF041853.1 - 1 255
AK295089.1 - 2 372
AK313359.1 - 2 382
AM177178.1 - 1 255
BC020890.1 - 2 379
BC032599.1 - 2 397
BC039592.1 - 2 371
BC045542.1 - 2 408 5298675
BI561509.1 - 2 414
HY154618.1 - 2 414

View Full Product Details