Gene Transcripts
- Gene Symbol
- FMO3
- Entrez Gene ID
- 2328
- Gene Name
- flavin containing monooxygenase 3
- Gene Aliases
- FMOII, TMAU, dJ127D3.1
- Chromosome Location
- Chr. 1: 171090873 - 171117819 on Build GRCh38
- UniGene ID
- Hs.445350
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001002294.2 |
NP_001002294.1
|
8, 9 |
1367 |
|
NM_001319173.1 |
NP_001306102.1
|
9, 10 |
1477 |
|
NM_001319174.1 |
NP_001306103.1
|
7, 8 |
1178 |
|
NM_006894.5 |
NP_008825.4
|
8, 9 |
1350 |
|
XM_011509345.2 |
XP_011507647.1
|
9, 10 |
1498 |
|
GenBank mRNA |
AK223166.1 |
- |
8, 9 |
1359 |
|
AK296105.1 |
- |
7, 8 |
1152 |
|
AK298406.1 |
- |
9, 10 |
1462 |
|
AK313197.1 |
- |
8, 9 |
1340 |
|
BC032016.1 |
- |
8, 9 |
1302 |
5175615 |
KU178077.1 |
- |
7, 8 |
1256 |
|
KU178078.1 |
- |
4, 5 |
366 |
|
Z47552.1 |
- |
8, 9 |
1349 |
|
View Full Product Details