Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005482.2 | NP_005473.1 | 5 | 474 | |
GenBank mRNA | AF022913.1 | - | 5 | 446 | |
AK304340.1 | - | 3 | 229 | ||
AK313015.1 | - | 5 | 457 | ||
BC020737.1 | - | 5 | 453 | 4722399 | |
BX648473.1 | - | 2 | 176 | ||
DA263434.1 | - | 5 | 474 | ||
Y07596.1 | - | 5 | 433 |