Gene Transcripts
- Gene Symbol
- PSMA2
- Entrez Gene ID
- 5683
- Gene Name
- proteasome subunit alpha 2
- Gene Aliases
- HC3, MU, PMSA2, PSC2
- Chromosome Location
- Chr. 7: 42916861 - 42932206 on Build GRCh38
- UniGene ID
- Hs.333786
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002787.4 |
NP_002778.1
|
4, 5 |
429 |
|
GenBank mRNA |
AK222976.1 |
- |
4, 5 |
397 |
|
AK290654.1 |
- |
4, 5 |
404 |
|
AU120930.1 |
- |
4, 5 |
404 |
|
BC002900.2 |
- |
3, 4 |
355 |
|
BC047697.1 |
- |
4, 5 |
383 |
5786518 |
BX641097.1 |
- |
3, 4 |
320 |
|
CB990982.1 |
- |
4, 5 |
472 |
|
CR450317.1 |
- |
4, 5 |
381 |
|
D00760.1 |
- |
4, 5 |
381 |
|
View Full Product Details