Gene Transcripts

Gene Symbol
GUCY1B2
Entrez Gene ID
2974
Gene Name
guanylate cyclase 1 soluble subunit beta 2 (pseudogene)
Gene Aliases
GC-SB2
Chromosome Location
Chr. 13: 50994511 - 51066157 on Build GRCh38
UniGene ID
Hs.411573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003923.2 5 554
GenBank mRNA AF038499.2 - 5 403
AK296746.1 - 6 710

View Full Product Details