Gene Transcripts
- Gene Symbol
- SLC35C2
- Entrez Gene ID
- 51006
- Gene Name
- solute carrier family 35 member C2
- Gene Aliases
- BA394O2.1, C20orf5, CGI-15, OVCOV1
- Chromosome Location
- Chr. 20: 46349528 - 46364461 on Build GRCh38
- UniGene ID
- Hs.593344
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281457.1 |
NP_001268386.1
|
4, 5 |
608 |
|
NM_001281458.1 |
NP_001268387.1
|
5, 6 |
547 |
|
NM_001281459.1 |
NP_001268388.1
|
4, 5 |
608 |
|
NM_001281460.1 |
NP_001268389.1
|
5, 6 |
1011 |
|
NM_015945.11 |
NP_057029.8
|
4, 5 |
679 |
|
NM_173073.3 |
NP_775096.1
|
4, 5 |
679 |
|
NM_173179.3 |
NP_775271.1
|
4, 5 |
874 |
|
XM_011528831.1 |
XP_011527133.1
|
4, 5 |
857 |
|
XM_011528832.1 |
XP_011527134.1
|
4, 5 |
1151 |
|
XM_011528833.1 |
XP_011527135.1
|
4, 5 |
675 |
|
XM_011528834.1 |
XP_011527136.1
|
4, 5 |
679 |
|
XM_011528836.1 |
XP_011527138.1
|
4, 5 |
760 |
|
XM_011528837.1 |
XP_011527139.1
|
4, 5 |
594 |
|
XM_017027856.1 |
XP_016883345.1
|
4, 5 |
1574 |
|
XM_017027857.1 |
XP_016883346.1
|
4, 5 |
1102 |
|
XM_017027858.1 |
XP_016883347.1
|
4, 5 |
1012 |
|
XM_017027859.1 |
XP_016883348.1
|
4, 5 |
1379 |
|
XM_017027860.1 |
XP_016883349.1
|
5, 6 |
819 |
|
XM_017027861.1 |
XP_016883350.1
|
4, 5 |
841 |
|
XM_017027862.1 |
XP_016883351.1
|
4, 5 |
679 |
|
GenBank mRNA |
AF132949.1 |
- |
4, 5 |
616 |
|
AF455052.1 |
- |
4, 5 |
634 |
|
AK222928.1 |
- |
4, 5 |
619 |
|
AK300207.1 |
- |
4, 5 |
562 |
|
AK300798.1 |
- |
4, 5 |
540 |
|
AK301022.1 |
- |
5, 6 |
489 |
|
AK315726.1 |
- |
4, 5 |
841 |
|
BC014191.2 |
- |
4, 5 |
623 |
4761663 |
BC021138.1 |
- |
4, 5 |
817 |
4870715 |
BC025277.1 |
- |
4, 5 |
875 |
4865888 |
BI831689.1 |
- |
5, 6 |
552 |
|
View Full Product Details