Gene Transcripts

Gene Symbol
UBE2D2
Entrez Gene ID
7322
Gene Name
ubiquitin conjugating enzyme E2 D2
Gene Aliases
E2(17)KB2, PUBC1, UBC4, UBC4/5, UBCH4, UBCH5B
Chromosome Location
Chr. 5: 139561166 - 139628434 on Build GRCh38
UniGene ID
Hs.108332
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003339.2 NP_003330.1 7 2488
NM_181838.1 NP_862821.1 8 2652
GenBank mRNA AF317220.1 - 5 1784
AK001428.1 - 7 2315

View Full Product Details