Gene Transcripts

Gene Symbol
RASL12
Entrez Gene ID
51285
Gene Name
RAS like family 12
Gene Aliases
RIS
Chromosome Location
Chr. 15: 65045576 - 65076654 on Build GRCh38
UniGene ID
Hs.27018
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001307930.1 NP_001294859.1 4 945
NM_016563.3 NP_057647.1 5 1002
XM_011521660.2 XP_011519962.1 6 815
GenBank mRNA AF233588.1 - 5 841
AK296261.1 - 4 755
AK300536.1 - 5 785
AK314914.1 - 5 810
BC053734.1 - 5 850 5588236

View Full Product Details