Gene Transcripts

Gene Symbol
RBP1
Entrez Gene ID
5947
Gene Name
retinol binding protein 1
Gene Aliases
CRABP-I, CRBP, CRBP1, CRBPI, RBPC
Chromosome Location
Chr. 3: 139517434 - 139539829 on Build GRCh38
UniGene ID
Hs.529571
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130992.1 NP_001124464.1 2 466
NM_001130993.1 NP_001124465.1 2 466
NM_002899.3 NP_002890.2 2 466
GenBank mRNA AK290315.1 - 2 273
AK301684.1 - 2 356
AK307378.1 - 2 275
AK309492.1 - 2 466
BC121052.2 - 2 224 40118103
CR541979.1 - 2 169
CR542005.1 - 2 169
DC356836.1 - 2 466
DC394326.1 - 2 356
M11433.1 - 2 294
M36809.1 - 2 241

View Full Product Details