Gene Transcripts

Gene Symbol
OSBPL11
Entrez Gene ID
114885
Gene Name
oxysterol binding protein like 11
Gene Aliases
ORP-11, ORP11, OSBP12, TCCCIA00292
Chromosome Location
Chr. 3: 125528858 - 125595537 on Build GRCh38
UniGene ID
Hs.477440
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022776.4 NP_073613.2 5 1365
GenBank mRNA AF346292.1 - 5 792
AF392454.1 - 5 936
AK292702.1 - 5 717
BC065213.1 - 5 715 3916115

View Full Product Details