Gene Transcripts

Gene Symbol
RNASET2
Entrez Gene ID
8635
Gene Name
ribonuclease T2
Gene Aliases
RNASE6PL, bA514O12.3
Chromosome Location
Chr. 6: 166929509 - 166956589 on Build GRCh38
UniGene ID
Hs.529989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003730.4 NP_003721.2 9 1220
XM_017011397.1 XP_016866886.1 10 999
XM_017011398.1 XP_016866887.1 8 1121
XM_017011399.1 XP_016866888.1 8 1135
GenBank mRNA AJ419865.1 - 9 1174
AK124363.1 - 2 2252
BC001660.2 - 9 1146 2959583
BC001819.2 - 9 1146 2959583
BC036282.1 - 9 1207
BC039713.1 - 9 1149 5225327
BC051912.1 - 9 1159 5227463
U85625.2 - 9 1177

View Full Product Details