Gene Transcripts

Gene Symbol
ULK3
Entrez Gene ID
25989
Gene Name
unc-51 like kinase 3
Gene Aliases
Chromosome Location
Chr. 15: 74836116 - 74843211 on Build GRCh38
UniGene ID
Hs.513034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099436.3 NP_001092906.3 4 505
NM_001284364.2 NP_001271293.2 4 505
NM_001284365.2 NP_001271294.1 4 428
NR_104307.2 4 505
NR_104308.2 4 505
NR_104309.2 4 505
XM_005254289.2 XP_005254346.1 5 520
XM_017022068.1 XP_016877557.1 5 554
XM_017022069.1 XP_016877558.1 5 558
XM_017022070.1 XP_016877559.1 5 571
XM_017022071.1 XP_016877560.1 5 558
XM_017022072.1 XP_016877561.1 4 734
GenBank mRNA AK075047.1 - 4 414
AK293181.1 - 4 358
AK293650.1 - 3 523
AK294241.1 - 4 439
AK294245.1 - 4 435
AK294251.1 - 4 439
AK298879.1 - 4 362
AK299380.1 - 4 432
AK316102.1 - 4 439
AL117482.1 - 4 640
BC036117.1 - 4 476
BC048289.1 - 4 433
BC056423.1 - 4 423
BC157884.1 - 4 487 8862706
BI829133.1 - 3 589
BQ927030.1 - 4 393

View Full Product Details