Gene Transcripts

Gene Symbol
UBXN1
Entrez Gene ID
51035
Gene Name
UBX domain protein 1
Gene Aliases
2B28, SAKS1, UBXD10
Chromosome Location
Chr. 11: 62676498 - 62679117 on Build GRCh38
UniGene ID
Hs.351296
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286077.1 NP_001273006.1 5, 6 670
NM_001286078.1 NP_001273007.1 3, 4 493
NM_015853.4 NP_056937.2 5, 6 670
XM_005274033.4 XP_005274090.1 6, 7 583
XM_011545090.2 XP_011543392.1 5, 6 629
XM_017017874.1 XP_016873363.1 6, 7 579
GenBank mRNA AK054832.1 - 4, 5 747
AK297856.1 - 5, 6 648
AK303471.1 - 4, 5 529
AK316012.1 - 5, 6 620
BC000902.2 - 5, 6 609 2900493
BC001372.2 - 5, 6 608 3051213
BC032689.1 - 4, 5 732
BC040129.1 - 5, 6 645 6065505
BI855821.1 - 3, 4 436
M68864.1 - 5, 6 614

View Full Product Details