Gene Transcripts
- Gene Symbol
- FAM63B
- Entrez Gene ID
- 54629
- Gene Name
- family with sequence similarity 63 member B
- Gene Aliases
- Chromosome Location
- Chr. 15: 58771192 - 58861900 on Build GRCh38
- UniGene ID
- Hs.591122
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001040450.2 |
NP_001035540.1
|
2, 3 |
1094 |
|
NM_001040453.2 |
NP_001035543.1
|
2, 3 |
924 |
|
XM_011521687.1 |
XP_011519989.1
|
2, 3 |
1092 |
|
XM_011521688.1 |
XP_011519990.1
|
2, 3 |
1092 |
|
XM_011521689.1 |
XP_011519991.1
|
2, 3 |
1092 |
|
XM_011521690.1 |
XP_011519992.1
|
2, 3 |
1092 |
|
XM_017022332.1 |
XP_016877821.1
|
2, 3 |
1092 |
|
XM_017022333.1 |
XP_016877822.1
|
2, 3 |
1092 |
|
XM_017022334.1 |
XP_016877823.1
|
2, 3 |
1092 |
|
XM_017022335.1 |
XP_016877824.1
|
2, 3 |
1092 |
|
GenBank mRNA |
AB032990.1 |
- |
2, 3 |
201 |
|
AK075319.1 |
- |
2, 3 |
972 |
|
AK297291.1 |
- |
2, 3 |
972 |
|
AL833610.1 |
- |
2, 3 |
1095 |
|
BC140811.1 |
- |
2, 3 |
924 |
9021679 |
BC144616.1 |
- |
2, 3 |
924 |
9053147 |
View Full Product Details