Gene Transcripts
- Gene Symbol
- SPEN
- Entrez Gene ID
- 23013
- Gene Name
- spen family transcriptional repressor
- Gene Aliases
- HIAA0929, MINT, RBM15C, SHARP
- Chromosome Location
- Chr. 1: 15847864 - 15940455 on Build GRCh38
- UniGene ID
- Hs.744843
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015001.2 |
NP_055816.2
|
1, 2 |
284 |
|
GenBank mRNA |
AF356524.1 |
- |
1, 2 |
284 |
|
View Full Product Details