Gene Transcripts

Gene Symbol
SWAP70
Entrez Gene ID
23075
Gene Name
SWAP switching B-cell complex 70kDa subunit
Gene Aliases
HSPC321, SWAP-70
Chromosome Location
Chr. 11: 9664077 - 9752991 on Build GRCh38
UniGene ID
Hs.153026
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297714.1 NP_001284643.1 11 1715
NM_015055.3 NP_055870.2 12 1889
XM_005252830.4 XP_005252887.1 10 1790
GenBank mRNA AB014540.1 - 12 1841
AF134894.1 - 12 1860
AK027131.1 - 9 1316
AK096859.1 - 11 1715
AK304104.1 - 12 1753
BC000616.2 - 12 1862 3346240

View Full Product Details