Gene Transcripts

Gene Symbol
MSTO1
Entrez Gene ID
55154
Gene Name
misato 1, mitochondrial distribution and morphology regulator
Gene Aliases
LST005, MST
Chromosome Location
Chr. 1: 155610170 - 155614967 on Build GRCh38
UniGene ID
Hs.656547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256532.1 NP_001243461.1 3 317
NM_001256533.1 NP_001243462.1 3 317
NM_018116.3 NP_060586.2 3 317
NR_046292.1 3 317
NR_046293.1 3 317
NR_046294.1 2 409
NR_046295.1 3 317
XM_006711421.1 XP_006711484.1 2 154
XM_006711423.2 XP_006711486.1 3 261
XM_011509695.1 XP_011507997.1 3 275
XM_011509696.1 XP_011507998.1 3 275
XM_011509697.1 XP_011507999.1 3 275
XM_011509698.1 XP_011508000.1 3 275
XM_011509699.1 XP_011508001.1 3 275
XM_011509700.1 XP_011508002.1 3 275
XM_011509701.1 XP_011508003.1 3 275
XM_011509702.1 XP_011508004.1 3 275
XM_011509703.1 XP_011508005.1 2 795
XM_011509704.1 XP_011508006.1 3 718
XM_011509705.1 XP_011508007.1 2 732
XM_011509706.1 XP_011508008.1 3 249
XM_011509707.1 XP_011508009.1 3 718
XM_011509708.1 XP_011508010.1 3 718
XM_011509709.2 XP_011508011.1 3 275
XM_017001603.1 XP_016857092.1 3 249
XM_017001604.1 XP_016857093.1 3 251
XM_017001605.1 XP_016857094.1 3 718
XM_017001606.1 XP_016857095.1 3 718
XM_017001607.1 XP_016857096.1 3 718
XM_017001608.1 XP_016857097.1 2 165
XM_017001609.1 XP_016857098.1 3 261
XM_017001610.1 XP_016857099.1 3 251
XM_017001611.1 XP_016857100.1 3 718
XM_017001612.1 XP_016857101.1 2 154
XM_017001613.1 XP_016857102.1 2 154
XM_017001614.1 XP_016857103.1 3 261
XM_017001615.1 XP_016857104.1 3 261
GenBank mRNA AF272833.1 - 3 195
AK001366.1 - 3 246
AK056128.1 - 2 338
AK092950.1 - 3 246
AK222863.1 - 3 243
AK290610.1 - 3 292
AK297140.1 - 3 246
AK309573.1 - 3 238
AY334564.1 - 2 360
BC002535.2 - 3 262 3140498
BC070067.1 - 3 269
BI911005.1 - 3 279
BX537684.1 - 3 269
CR749791.1 - 3 243
CR936872.1 - 2 766

Gene Symbol
MSTO2P
Entrez Gene ID
100129405
Gene Name
misato family member 2, pseudogene
Gene Aliases
MSTO2
Chromosome Location
Chr. 1: 155745768 - 155750688 on Build GRCh38
UniGene ID
Hs.656361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024117.2 3 299
GenBank mRNA AK293928.1 - 3 299
AK295146.1 - 3 299
BC099909.1 - 3 249
DC387604.1 - 3 299

View Full Product Details