Gene Transcripts

Gene Symbol
FAM213A
Entrez Gene ID
84293
Gene Name
family with sequence similarity 213 member A
Gene Aliases
C10orf58, PAMM
Chromosome Location
Chr. 10: 80407829 - 80437115 on Build GRCh38
UniGene ID
Hs.500333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243778.1 NP_001230707.1 6 781
NM_001243779.1 NP_001230708.1 6 851
NM_001243780.1 NP_001230709.1 6 1347
NM_001243781.1 NP_001230710.1 6 864
NM_001243782.1 NP_001230711.1 6 764
NM_032333.4 NP_115709.3 6 881
XM_011540265.2 XP_011538567.1 6 1184
XM_011540266.2 XP_011538568.1 6 1587
XM_017016775.1 XP_016872264.1 6 868
GenBank mRNA AF086462.1 - 127
AF305824.1 - 4 537
AK074643.1 - 6 826
AK075447.1 - 6 856
AY359059.1 - 6 774
BC005871.2 - 6 743 3010078

View Full Product Details