Gene Transcripts

Gene Symbol
ALG14
Entrez Gene ID
199857
Gene Name
ALG14, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CMS15
Chromosome Location
Chr. 1: 94981565 - 95072974 on Build GRCh38
UniGene ID
Hs.408927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305242.1 NP_001292171.1 2 254
NM_144988.3 NP_659425.1 2 254
XM_005270582.3 XP_005270639.1 2 276
XM_011540897.2 XP_011539199.1 2 276
GenBank mRNA AK289395.1 - 2 254
AW242346.1 - 2 168
BC011706.2 - 2 230 3689162

View Full Product Details