Gene Transcripts

Gene Symbol
KNCN
Entrez Gene ID
148930
Gene Name
kinocilin
Gene Aliases
Kino, L5
Chromosome Location
Chr. 1: 46545638 - 46551657 on Build GRCh38
UniGene ID
Hs.350764
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097611.1 NP_001091080.1 3 910
NM_001322255.1 NP_001309184.1 4 1421
GenBank mRNA AK056573.1 - 3 838
BC101295.1 - 3 832
BC101296.1 - 3 832
BC101297.2 - 3 832
BC101298.2 - 3 832

View Full Product Details